Learn More
Medchemexpress LLC HY-101412 5mg Medchemexpress, 2-(Methylamino)-1H-purin-6(7H)-one CAS:10030-78-1 Purity:>98%
SDP
Catalog No. 501932116
Encompass_Preferred
Medchemexpress, HY-101412 5mg 2-(Methylamino)-1H-purin-6(7H)-one CAS:10030-78-1 2-(Methylamino)-1H-purin-6(7H)-one (N2-Methylguanine) is a modified nucleoside. 2-(Methylamino)-1H-purin-6(7H)-one is an endogenous methylated nucleoside found in human fluids. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2116 Supplier Medchemexpress LLC Supplier No. HY1014125MG
Medchemexpress, HY-101412 5mg 2-(Methylamino)-1H-purin-6(7H)-one CAS:10030-78-1 2-(Methylamino)-1H-purin-6(7H)-one (N2-Methylguanine) is a modified nucleoside. 2-(Methylamino)-1H-purin-6(7H)-one is an endogenous methylated nucleoside found in human fluids. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.