Learn More
Medchemexpress LLC 2-(methylamino)-1H-purin-6(7H)-one (N2-methylguanine) | 10030-78-1 | MFCD00127896 | 98.0% | 165.15 g/mol | C6H7N5O | 100 MG
SDP
Catalog No. 5000247026
Encompass_Preferred
2-(Methylamino)-1H-purin-6(7H)-one (N2-methylguanine) is a modified purine nucleobase and endogenous methylated nucleoside supplied as a solid research reagent. It is used as a reference standard and analytical reagent in studies of nucleic acid modification, enzymology, and biomarker analysis.
  • Modified purine nucleobase relevant to nucleic acid research.
  • High purity suitable for analytical and biochemical assays.
  • Available in multiple milligram-scale sizes for experimental flexibility.
  • Solid physical form for straightforward handling and storage.
  • Suitable as a reference standard for modification and biomarker studies.
Catalog No. 50-002-47026 Supplier Medchemexpress LLC Supplier No. HY101412100MG
2-(Methylamino)-1H-purin-6(7H)-one (N2-methylguanine) is a modified purine nucleobase and endogenous methylated nucleoside supplied as a solid research reagent. It is used as a reference standard and analytical reagent in studies of nucleic acid modification, enzymology, and biomarker analysis.
  • Modified purine nucleobase relevant to nucleic acid research.
  • High purity suitable for analytical and biochemical assays.
  • Available in multiple milligram-scale sizes for experimental flexibility.
  • Solid physical form for straightforward handling and storage.
  • Suitable as a reference standard for modification and biomarker studies.