Learn More
Medchemexpress LLC 2-(Methylamino)acetic acid-d3 hydrochloride | 347840-04-4 | 98.6% | 128.57 | 10 MG
SDP
Catalog No. 5000454842
Encompass_Preferred
This product is 2-(Methylamino)acetic acid-d3 hydrochloride, a deuterated derivative of methylaminoacetic acid. It is supplied as a white to off-white solid with a high purity of 98.60% as determined by HPLC, and an isotopic enrichment of 99.9%. It is intended for laboratory use, and its properties are confirmed through a Certificate of Analysis. The molecular formula is C3H5D3ClNO2 with a molecular weight of 128.57. It requires sealed storage at 4°C, away from moisture, or -80°C/-20°C when in solvent.
  • High purity (98.6%) confirmed by HPLC
  • High isotopic enrichment (99.9%)
  • Supplied as a solid for various laboratory applications
  • Requires specific storage conditions to maintain stability
Catalog No. 50-004-54842 Supplier Medchemexpress LLC Supplier No. HYW012791S10MG
This product is 2-(Methylamino)acetic acid-d3 hydrochloride, a deuterated derivative of methylaminoacetic acid. It is supplied as a white to off-white solid with a high purity of 98.60% as determined by HPLC, and an isotopic enrichment of 99.9%. It is intended for laboratory use, and its properties are confirmed through a Certificate of Analysis. The molecular formula is C3H5D3ClNO2 with a molecular weight of 128.57. It requires sealed storage at 4°C, away from moisture, or -80°C/-20°C when in solvent.
  • High purity (98.6%) confirmed by HPLC
  • High isotopic enrichment (99.9%)
  • Supplied as a solid for various laboratory applications
  • Requires specific storage conditions to maintain stability