Learn More
Medchemexpress LLC 2-(Methylamino)acetic acid hydrochloride | 637-96-7 | ≥98.0% | 125.55 | 100 G
SDP
Catalog No. 5000454921
Encompass_Preferred
2-(Methylamino)acetic acid hydrochloride is a derivative of Glycine. This compound is used commercially as an ergogenic supplement, known for its ability to influence the secretion of anabolic hormones, supply fuel during exercise, and enhance mental performance during stress-related tasks. It also helps prevent exercise-induced muscle damage.
  • Derived from Glycine.
  • Functions as an ergogenic supplement.
  • Influences anabolic hormone secretion.
  • Provides fuel during physical activity.
  • Enhances mental performance under stress.
  • Helps prevent exercise-induced muscle damage.
  • Solid, white to off-white appearance.
  • Shipping at room temperature.
  • Recommended storage at 4°C, sealed, away from moisture.
  • In solvent, stores at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-54921 Supplier Medchemexpress LLC Supplier No. HYW012791100G
2-(Methylamino)acetic acid hydrochloride is a derivative of Glycine. This compound is used commercially as an ergogenic supplement, known for its ability to influence the secretion of anabolic hormones, supply fuel during exercise, and enhance mental performance during stress-related tasks. It also helps prevent exercise-induced muscle damage.
  • Derived from Glycine.
  • Functions as an ergogenic supplement.
  • Influences anabolic hormone secretion.
  • Provides fuel during physical activity.
  • Enhances mental performance under stress.
  • Helps prevent exercise-induced muscle damage.
  • Solid, white to off-white appearance.
  • Shipping at room temperature.
  • Recommended storage at 4°C, sealed, away from moisture.
  • In solvent, stores at -80°C for 6 months or -20°C for 1 month.