Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(methylamino)benzoic acid | 119-68-6 | 99.9% | 151.16 g/mol | C8H9NO2 | 10 G
SDP

Catalog No. 5000208733
Encompass_Preferred

2-(methylamino)benzoic acid is a small-molecule organic compound supplied as a high-purity solid for research and analytical use. It is a metabolite of methyl N-methylanthranilates and may be used as a reference standard or synthetic intermediate.

  • High purity: 99.9%.
  • Molecular weight: 151.16 g/mol.
  • Chemical formula: C8H9NO2.
  • Available as a 10 G solid for laboratory use.

Catalog No. 50-002-08733 Supplier Medchemexpress LLC Supplier No. HYW04070510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(methylamino)benzoic acid is a small-molecule organic compound supplied as a high-purity solid for research and analytical use. It is a metabolite of methyl N-methylanthranilates and may be used as a reference standard or synthetic intermediate.

  • High purity: 99.9%.
  • Molecular weight: 151.16 g/mol.
  • Chemical formula: C8H9NO2.
  • Available as a 10 G solid for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.