Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(methylamino)benzoic acid | 119-68-6 | MFCD00002424 | 99.8% | 151.16 g/mol | C8H9NO2 | 1 ML
SDP

Catalog No. 5000209720
Encompass_Preferred

2-(methylamino)benzoic acid is a small-molecule research reagent (also known as N-methylanthranilic acid) provided for laboratory use. It is supplied as a solid and as a 10 mM solution in DMSO and is accompanied by a certificate of analysis and safety documentation.

  • Small-molecule reagent for research applications.
  • Available as 10 mM solution in DMSO and as solid forms.
  • High purity (HPLC 99.82%).
  • Molecular formula C8H9NO2 and molecular weight 151.16 g/mol.
  • CAS number 119-68-6 for unambiguous identification.

Catalog No. 50-002-09720 Supplier Medchemexpress LLC Supplier No. HYW04070510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(methylamino)benzoic acid is a small-molecule research reagent (also known as N-methylanthranilic acid) provided for laboratory use. It is supplied as a solid and as a 10 mM solution in DMSO and is accompanied by a certificate of analysis and safety documentation.

  • Small-molecule reagent for research applications.
  • Available as 10 mM solution in DMSO and as solid forms.
  • High purity (HPLC 99.82%).
  • Molecular formula C8H9NO2 and molecular weight 151.16 g/mol.
  • CAS number 119-68-6 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.