Learn More
Medchemexpress LLC 2-(methylamino)ethanol | 109-83-1 | MFCD00002839 | 99.5% | 500g
SDP
Catalog No. 5000266171
Encompass_Preferred
2-(Methylamino)ethanol (N-Methylethanolamine) is an alkaloid that can be isolated from lentil seeds 2-(Methylamino)ethanol is used as an organic building block in chemical synthesis and as a solvent for removing carbon dioxide from gas streams[1]
Catalog No. 50-002-66171 Supplier Medchemexpress LLC Supplier No. HYY1783500G
2-(Methylamino)ethanol (N-Methylethanolamine) is an alkaloid that can be isolated from lentil seeds 2-(Methylamino)ethanol is used as an organic building block in chemical synthesis and as a solvent for removing carbon dioxide from gas streams[1]