Learn More
Medchemexpress LLC 2-methylaminoethanol | 109-83-1 | MFCD00002839 | 1mL
SDP
Catalog No. 5000266173
Encompass_Preferred
2-(Methylamino)ethanol (N-Methylethanolamine) is an alkaloid that can be isolated from lentil seeds 2-(Methylamino)ethanol is used as an organic building block in chemical synthesis and as a solvent for removing carbon dioxide from gas streams[1]
Catalog No. 50-002-66173 Supplier Medchemexpress LLC Supplier No. HYY178310MM/1ML
2-(Methylamino)ethanol (N-Methylethanolamine) is an alkaloid that can be isolated from lentil seeds 2-(Methylamino)ethanol is used as an organic building block in chemical synthesis and as a solvent for removing carbon dioxide from gas streams[1]