Learn More
Medchemexpress LLC 2-Methylanisole | 578-58-5 | MFCD00008373 | 99.8% | 122.17 g/mol | C8H10O | 100 G
SDP
Catalog No. 5000300143
Encompass_Preferred
2-Methylanisole (1-methoxy-2-methylbenzene) is an organic monomethoxybenzene used as a chemical intermediate in organic synthesis, particularly for preparing compounds with a methylhydroquinone core. It is supplied for research use in liquid and solution forms, with manufacturer-specified purity and storage recommendations for both pure material and solutions.
  • Used as an intermediate in organic synthesis.
  • Chemical identifiers: CAS 578-58-5; formula C8H10O; molecular weight 122.17 g/mol.
  • High purity suitable for research (reported 99.0-99.8%).
  • Available in solution and bulk liquid pack sizes, including 100 g.
  • Supplier storage guidance provided for pure form and for material in solvent.
Catalog No. 50-003-00143 Supplier Medchemexpress LLC Supplier No. HYW027751100G
2-Methylanisole (1-methoxy-2-methylbenzene) is an organic monomethoxybenzene used as a chemical intermediate in organic synthesis, particularly for preparing compounds with a methylhydroquinone core. It is supplied for research use in liquid and solution forms, with manufacturer-specified purity and storage recommendations for both pure material and solutions.
  • Used as an intermediate in organic synthesis.
  • Chemical identifiers: CAS 578-58-5; formula C8H10O; molecular weight 122.17 g/mol.
  • High purity suitable for research (reported 99.0-99.8%).
  • Available in solution and bulk liquid pack sizes, including 100 g.
  • Supplier storage guidance provided for pure form and for material in solvent.