Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methylanisole | 578-58-5 | MFCD00008373 | 99.8% | 122.17 g/mol | C8H10O | 50 G
SDP

Catalog No. 5000299332
Encompass_Preferred

2-Methylanisole is a monomethoxybenzene used as a chemical intermediate in the preparation of compounds containing a methylhydroquinone core. It is supplied as a high-purity liquid in multiple pack sizes for research and synthetic applications.

  • High purity (99.8%).
  • Molecular weight 122.17 g/mol.
  • Molecular formula C8H10O.
  • Suitable as a chemical intermediate for organic synthesis.
  • Supplied as a liquid in multiple pack sizes, including 50 g.
  • Supporting documentation available: datasheet, SDS, COA.

Catalog No. 50-002-99332 Supplier Medchemexpress LLC Supplier No. HYW02775150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methylanisole is a monomethoxybenzene used as a chemical intermediate in the preparation of compounds containing a methylhydroquinone core. It is supplied as a high-purity liquid in multiple pack sizes for research and synthetic applications.

  • High purity (99.8%).
  • Molecular weight 122.17 g/mol.
  • Molecular formula C8H10O.
  • Suitable as a chemical intermediate for organic synthesis.
  • Supplied as a liquid in multiple pack sizes, including 50 g.
  • Supporting documentation available: datasheet, SDS, COA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.