Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methylanisole-d3 | 258832-47-2 | ≥98.0% | 125.18 g·mol⁻¹ | C8H7D3O | 100 MG
SDP

Catalog No. 5000301562
Encompass_Preferred

2-Methylanisole-d3 is the deuterium-labeled isotopologue of 2-methylanisole provided for laboratory research. It is used as an isotopic standard and as a synthetic intermediate in the preparation of compounds with a methylhydroquinone core. For research use only; not for human consumption.

  • Isotopic enrichment: 99 atom % D.
  • Molecular formula: C8H7D3O.
  • Molecular weight: 125.18 g·mol⁻¹.
  • Typical chemical purity: ≥98.0%.
  • Packaging: 100 mg suitable for analytical and synthetic workflows.
  • Intended use: research applications and method development.

Catalog No. 50-003-01562 Supplier Medchemexpress LLC Supplier No. HYW027751S100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methylanisole-d3 is the deuterium-labeled isotopologue of 2-methylanisole provided for laboratory research. It is used as an isotopic standard and as a synthetic intermediate in the preparation of compounds with a methylhydroquinone core. For research use only; not for human consumption.

  • Isotopic enrichment: 99 atom % D.
  • Molecular formula: C8H7D3O.
  • Molecular weight: 125.18 g·mol⁻¹.
  • Typical chemical purity: ≥98.0%.
  • Packaging: 100 mg suitable for analytical and synthetic workflows.
  • Intended use: research applications and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.