Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methylanisole-d3 | 258832-47-2 | ≥98.0% | 125.18 g·mol⁻¹ | C8H7D3O | 25 MG
SDP

Catalog No. 5000303192
Encompass_Preferred

2-Methylanisole-d3 is the deuterium-labeled analogue of 2-methylanisole supplied as a liquid isotopic reagent for synthetic chemistry and analytical applications. It is used as an internal standard in mass spectrometry and as a deuterated intermediate in the preparation of labeled compounds.

  • Contains three deuterium atoms for isotopic labeling (molecular formula C8H7D3O).
  • Molecular weight 125.18 g·mol⁻¹.
  • Useful as an internal standard in mass spectrometry and quantitative analyses.
  • Serves as a deuterated intermediate for synthesis of labeled compounds.
  • Appearance: light brown to brown liquid; supplier documentation reports isotopic enrichment and chemical purity.

Catalog No. 50-003-03192 Supplier Medchemexpress LLC Supplier No. 25MGHYW027751S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methylanisole-d3 is the deuterium-labeled analogue of 2-methylanisole supplied as a liquid isotopic reagent for synthetic chemistry and analytical applications. It is used as an internal standard in mass spectrometry and as a deuterated intermediate in the preparation of labeled compounds.

  • Contains three deuterium atoms for isotopic labeling (molecular formula C8H7D3O).
  • Molecular weight 125.18 g·mol⁻¹.
  • Useful as an internal standard in mass spectrometry and quantitative analyses.
  • Serves as a deuterated intermediate for synthesis of labeled compounds.
  • Appearance: light brown to brown liquid; supplier documentation reports isotopic enrichment and chemical purity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.