Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylanthraquinone 99.0+%, TCI America™
SDP

Catalog No. M015625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M015625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M015625G Supplier TCI America Supplier No. M015625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 84-54-8
Melting Point 175°C
Color Yellow
Molecular Formula C15H10O2
Quantity 25 g
MDL Number MFCD00001235
Synonym 2-methylanthraquinone, tectoquinone, 2-methyl anthraquinone, techtoquinone, tectochinon, 2-methyl-9,10-anthraquinone, 9,10-anthracenedione, 2-methyl, beta-methylanthraquinone, 2-methylanthra-9,10-quinone, anthraquinone, 2-methyl
InChI Key NJWGQARXZDRHCD-UHFFFAOYSA-N
SMILES CC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
IUPAC Name 2-methylanthracene-9,10-dione
Molecular Weight (g/mol) 222.243
PubChem CID 6773
ChEBI CHEBI:9427
Formula Weight 222.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylanthraquinone
Show More Show Less
EINECSNumber (4)-0687
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.