Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzhydrol 98.0+%, TCI America™
SDP

Catalog No. M130925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M130925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M130925G Supplier TCI America Supplier No. M130925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5472-13-9
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.265
MDL Number MFCD00004490
InChI Key MXHXXJOHFRHBFB-UHFFFAOYSA-N
Synonym 2-methylbenzhydrol, phenyl o-tolyl methanol, benzhydrol, 2-methyl, 2-methylphenyl phenylmethanol, o-methylbenzhydrol, phenyl-o-tolyl-methanol, 2-methylphenyl-phenylmethanol, benzenemethanol, 2-methyl-.alpha.-phenyl, 2-methylphenyl phenyl methanol, phenyl-o-tolylcarbinol
PubChem CID 95659
IUPAC Name (2-methylphenyl)-phenylmethanol
SMILES CC1=CC=CC=C1C(C2=CC=CC=C2)O

Specifications

Melting Point 92°C
Color White
Quantity 25 g
Formula Weight 198.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylbenzhydrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.