Learn More
Medchemexpress LLC 2-Methylbenzo[d]oxazol-6-ol | 5078-07-9 | 99.8% | C8H7NO2 | 1 G
SDP
Catalog No. 5000433991
Encompass_Preferred
2-Methylbenzo[d]oxazol-6-ol is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It is suitable for laboratory chemicals and the manufacture of substances.
  • Identified for laboratory use and substance manufacturing
  • Stable as a powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month
  • High purity of 99.84% (LCMS)
Catalog No. 50-004-33991 Supplier Medchemexpress LLC Supplier No. HY651721G
2-Methylbenzo[d]oxazol-6-ol is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It is suitable for laboratory chemicals and the manufacture of substances.
  • Identified for laboratory use and substance manufacturing
  • Stable as a powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month
  • High purity of 99.84% (LCMS)