Learn More
Medchemexpress LLC 2-Methylbenzo[d]oxazol-6-ol | 5078-07-9 | C8H7NO2 | 100 MG
SDP
Catalog No. 5000433789
Encompass_Preferred
2-Methylbenzo[d]oxazol-6-ol is a chemical compound with the formula C8H7NO2. It appears as a solid and is stable under recommended storage conditions. This product is intended for laboratory research and the manufacture of substances.
  • Stable under recommended storage conditions.
  • Solid appearance.
  • Used as a laboratory chemical.
  • Used in the manufacture of substances.
  • Recommended storage for powder at -20°C for 3 years, or 4°C for 2 years.
  • Recommended storage in solvent at -80°C for 6 months, or -20°C for 1 month.
Catalog No. 50-004-33789 Supplier Medchemexpress LLC Supplier No. HY65172100MG
2-Methylbenzo[d]oxazol-6-ol is a chemical compound with the formula C8H7NO2. It appears as a solid and is stable under recommended storage conditions. This product is intended for laboratory research and the manufacture of substances.
  • Stable under recommended storage conditions.
  • Solid appearance.
  • Used as a laboratory chemical.
  • Used in the manufacture of substances.
  • Recommended storage for powder at -20°C for 3 years, or 4°C for 2 years.
  • Recommended storage in solvent at -80°C for 6 months, or -20°C for 1 month.