Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbenzothiazol-6-ol, 96%

Catalog No. AAH3381203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3381203 1 g
AAH3381206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3381203 Supplier Thermo Scientific Chemicals Supplier No. H3381203
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 68867-18-5 | C8H7NOS | 165.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68867-18-5
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00227234
InChI Key ROFBPPIQUBJMRO-UHFFFAOYSA-N
Synonym 2-methyl-6-benzothiazolol, 2-methylbenzo d thiazol-6-ol, 6-benzothiazolol, 2-methyl, 2-methylbenzothiazol-6-ol, 6-hydroxy-2-methylbenzothiazole, 6-hydroxy-2-methylbenzthiazole, pubchem17329, 2-methyl-benzothiazol-6-ol, 2-methyl-6-hydroxybenzothiazole, 6-hydroxy-2-methyl-benzothiazole
PubChem CID 759304
IUPAC Name 2-methyl-1,3-benzothiazol-6-ol
SMILES CC1=NC2=CC=C(O)C=C2S1

Specifications

Melting Point 159°C to 161°C
Quantity 1 g
Formula Weight 165.21
Percent Purity 96%
Chemical Name or Material 2-Methylbenzothiazol-6-ol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.