Learn More
Medchemexpress LLC 2-methylbutanal (2-methylbutyraldehyde) | 96-17-3 | MFCD00006984 | 98.6% | 86.13 g·mol⁻¹ | C5H10O | 25 G
SDP
Catalog No. 5000302208
Encompass_Preferred
2-Methylbutanal (2-methylbutyraldehyde) is a small aliphatic aldehyde used as a volatile biomarker for detection of Proteus species and as a building block in organic synthesis and flavor/fragrance research. It is a colorless to light-yellow liquid with chemical formula C5H10O, molecular weight 86.13 g·mol⁻¹, and typical reported purity of about 98.6%.
  • Used as a volatile biomarker for detection of Proteus species.
  • High purity suitable for analytical and research applications.
  • Liquid physical form with defined density and molecular properties for formulation.
  • Useful building block in organic synthesis and flavor chemistry.
  • Available in small laboratory pack sizes for convenient research use.
Catalog No. 50-003-02208 Supplier Medchemexpress LLC Supplier No. 25GHYW020678
Add to Cart
Add to Cart
2-Methylbutanal (2-methylbutyraldehyde) is a small aliphatic aldehyde used as a volatile biomarker for detection of Proteus species and as a building block in organic synthesis and flavor/fragrance research. It is a colorless to light-yellow liquid with chemical formula C5H10O, molecular weight 86.13 g·mol⁻¹, and typical reported purity of about 98.6%.
  • Used as a volatile biomarker for detection of Proteus species.
  • High purity suitable for analytical and research applications.
  • Liquid physical form with defined density and molecular properties for formulation.
  • Useful building block in organic synthesis and flavor chemistry.
  • Available in small laboratory pack sizes for convenient research use.