Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Methylbutyl acetate 99 f1KG
Catalog No. 502899650
Encompass
2-Methylbutyl acetate is one of the key volatile aroma compounds that contribute to the characteristic aroma of apples of different varieties such as royal gala apple and fiji apples. It also occurs naturally in pear and banana.
Catalog No. 50-289-9650 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W3644011KGK
2-Methylbutyl acetate is one of the key volatile aroma compounds that contribute to the characteristic aroma of apples of different varieties such as royal gala apple and fiji apples. It also occurs naturally in pear and banana.