Learn More
Medchemexpress LLC 2-methylbutyl β-D-glucopyranoside | 80202-03-5 | >98.0% | 10 MG
SDP
Catalog No. 5000296510
Encompass_Preferred
2-Methylbutyl β-D-glucopyranoside is a glycoside found in apple fruit that functions as an aroma precursor; enzymatic hydrolysis releases volatile 2-methylbutanol and makes it useful for flavor chemistry research.
  • Acts as an aroma precursor in flavor chemistry studies.
  • High purity (>98%) suitable for analytical applications.
  • Soluble in DMSO and water for solution preparation.
  • Available in small laboratory pack sizes (1 mg, 5 mg, 10 mg) for research workflows.
Catalog No. 50-002-96510 Supplier Medchemexpress LLC Supplier No. HYN1224710MG
2-Methylbutyl β-D-glucopyranoside is a glycoside found in apple fruit that functions as an aroma precursor; enzymatic hydrolysis releases volatile 2-methylbutanol and makes it useful for flavor chemistry research.
  • Acts as an aroma precursor in flavor chemistry studies.
  • High purity (>98%) suitable for analytical applications.
  • Soluble in DMSO and water for solution preparation.
  • Available in small laboratory pack sizes (1 mg, 5 mg, 10 mg) for research workflows.