Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbutyl DL-2-Methylbutyrate 97.0+%, TCI America™
SDP

Catalog No. M057825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M057825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M057825ML Supplier TCI America Supplier No. M057825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2445-78-5
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.268
MDL Number MFCD00059395
InChI Key PVYFCGRBIREQLL-UHFFFAOYSA-N
Synonym DL-2-Methylbutyric Acid 2-Methylbutyl Ester
PubChem CID 17129
IUPAC Name 2-methylbutyl 2-methylbutanoate
SMILES CCC(C)COC(=O)C(C)CC

Specifications

Color Yellow
Boiling Point 187°C
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbutyl DL-2-Methylbutyrate
EINECSNumber (2)-0776
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.