Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-2-Methylbutyl-p-aminocinnamate 98%, Thermo Scientific™

Catalog No. AC414100050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC414100050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC414100050 Supplier Thermo Scientific Chemicals Supplier No. 414100050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62742-50-1
Molecular Formula C14H19NO2
Molecular Weight (g/mol) 233.311
InChI Key OUZTVIGSXSIPAZ-OALRPVAKSA-N
Synonym 2r-2-methylbutyl 2e-3-4-aminophenyl prop-2-enoate, 2-propenoic acid, 3-4-aminophenyl-, 2r-2-methylbutyl ester, 2e, r,e-2-methylbutyl 3-4-aminophenyl acrylate, 2r-2-methylbutyl e-3-4-aminophenyl prop-2-enoate, 3-4-aminophenyl propenoic acid r-2-methylbutyl ester, 2e-3-4-aminophenyl-2-propenoic acid 2r-2-methylbutyl ester
PubChem CID 5702745
IUPAC Name [(2R)-2-methylbutyl] (E)-3-(4-aminophenyl)prop-2-enoate
SMILES CCC(C)COC(=O)C=CC1=CC=C(C=C1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Quantity 5 g
Formula Weight 233.31
Chemical Name or Material (+)-2-Methylbutyl-p-aminocinnamate, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.