Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbutyraldehyde, 95%

Catalog No. aaa1965022
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1965022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1965022 Supplier Thermo Scientific Chemicals Supplier No. A1965022
Only null left
Add to Cart

CAS: 96-17-3 | C5H10O | 86.13 g/mol

2-Methylbutyraldehyde is used for lift in woody and cocoa topnote complexes. And also used in cocoa, coffee, wine and nut flavors for lift.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbutyraldehyde is used for lift in woody and cocoa topnote complexes. And also used in cocoa, coffee, wine and nut flavors for lift.

Solubility
Soluble in water, ether, and alcohol.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-17-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006984
InChI Key BYGQBDHUGHBGMD-UHFFFAOYNA-N
Synonym 2-methylbutyraldehyde, butanal, 2-methyl, 2-formylbutane, 2-methylbutyric aldehyde, methylethylacetaldehyde, 2-ethylpropanal, 2-methyl-1-butanal, butyraldehyde, 2-methyl, acetaldehyde, ethylmethyl, alpha-methylbutanal
PubChem CID 7284
ChEBI CHEBI:16182
IUPAC Name 2-methylbutanal
SMILES CCC(C)C=O

Specifications

Density 0.806
Boiling Point 94°C to 96°C
Flash Point 4°C (39°F)
Refractive Index 1.3919
Quantity 100 g
UN Number UN1989
Beilstein 1633540
Sensitivity Air Sensitive
Solubility Information Soluble in water,ether,and alcohol.
Formula Weight 86.13
Percent Purity 95%
Chemical Name or Material 2-Methylbutyraldehyde
Show More Show Less
Hazard Category H225-H317-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 202-485-6
RTECSNumber ES3400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.