Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbutyric acid, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020063 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0063 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0063 Supplier MilliporeSigma Supelco Supplier No. 496591ML
Add to Cart
Add to Cart

Specifications

CAS 116-53-0
Vapor Pressure 0.5 mmHg (20°C)
Density 0.936 g/mL (at 25°C)
Flash Point 74°C - Closed Cup
Linear Formula C2H5 CH(CH3)CO2 H
MDL Number MFCD00002669
Boiling Point 176°C to 177°C
Synonym NSC 7304
Type Analytical Standard
Quantity 1 mL
Formula Weight 102.13
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.