Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbutyronitrile 95.0+%, TCI America™
SDP

Catalog No. M070625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M070625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M070625ML Supplier TCI America Supplier No. M070625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18936-17-9
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00001877
InChI Key RCEJCSULJQNRQQ-UHFFFAOYSA-N
Synonym sec-Butyl Cyanide, 2-Cyanobutane
PubChem CID 29339
ChEBI CHEBI:61669
IUPAC Name 2-methylbutanenitrile
SMILES CCC(C)C#N

Specifications

Color Yellow
Boiling Point 125°C
Quantity 25 mL
UN Number 3275
Formula Weight 83.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbutyronitrile
RTECSNumber EU0530000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.