Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylbutyryl chloride, 99%

Catalog No. aab2259906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2259906 5 g
AAB2259914 25 g
AAB2259922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2259906 Supplier Thermo Scientific Chemicals Supplier No. B2259906
Only null left
Add to Cart

CAS: 57526-28-0 | C5H9ClO | 120.576 g/mol

2-Methylbutyryl chloride was used in the synthesis of 2-methylbutyramide, N-((R,S)-2-methylbutanoyl)salicylhydrazide, 2-methyl-1-phenyl-1-butanone, 1,1’-(1,3,7,9-tetrahydroxydibenzofuran-2,6-diyl)bis(2-methylbutan-1-one).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbutyryl chloride was used in the synthesis of 2-methylbutyramide, N-((R,S)-2-methylbutanoyl)salicylhydrazide, 2-methyl-1-phenyl-1-butanone, 1,1’-(1,3,7,9-tetrahydroxydibenzofuran-2,6-diyl)bis(2-methylbutan-1-one).

Solubility
Reacts with water.

Notes
Store away from oxidizing agents, amines, bases, alcohols, active metals, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57526-28-0
Molecular Formula C5H9ClO
Molecular Weight (g/mol) 120.576
MDL Number MFCD00041726
InChI Key XRPVXVRWIDOORM-UHFFFAOYSA-N
Synonym 2-methylbutyryl chloride, dl-2-methylbutyryl chloride, butanoyl chloride, 2-methyl, ethylmethylacetylchloride, dl-2-methyl-butyryl chloride, rs-2-methylbutyryl chloride, 2-methyl-butyryl chloride, #, +-2-methyl butanoyl chloride
PubChem CID 93697
IUPAC Name 2-methylbutanoyl chloride
SMILES CCC(C)C(=O)Cl

Specifications

Density 0.98
Boiling Point 116°C to 117°C
Flash Point 20°C (68°F)
Refractive Index 1.418
Quantity 5 g
UN Number UN2924
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 120.58
Percent Purity 99%
Chemical Name or Material 2-Methylbutyryl chloride
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 260-787-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.