Learn More
Medchemexpress LLC 2-methylbutyryl-L-carnitine-d3 (chloride) | 00-00-0 | MFCD34676379 | 99.2% | 284.79 g/mol | C12H21D3ClNO4 | 5 MG
SDP
Catalog No. 5000262225
Encompass_Preferred
2-Methylbutyryl-L-carnitine-d3 (chloride) is the deuterium-labeled form of 2-methylbutyryl-L-carnitine supplied as the chloride salt. It is intended for use as a stable isotope internal standard or tracer in analytical chemistry and biochemical assays, particularly for LC-MS quantitation.
Catalog No. 50-002-62225 Supplier Medchemexpress LLC Supplier No. HY132958S5MG
2-Methylbutyryl-L-carnitine-d3 (chloride) is the deuterium-labeled form of 2-methylbutyryl-L-carnitine supplied as the chloride salt. It is intended for use as a stable isotope internal standard or tracer in analytical chemistry and biochemical assays, particularly for LC-MS quantitation.