Learn More
Cayman Chemical 2-MethylcItrIc AcId 10mg
SDP
Catalog No. 502988009
Encompass_Preferred
An endogenous tricarboxylic acid; formed by the condensation of propionyl-CoA with oxaloacetic acid by citrate synthase under conditions of propionyl-CoA accumulation; accumulation is associated with cobalamin deficiencies, propionic acidemia, and methylmalonic acidurias
Catalog No. 50-298-8009 Supplier Cayman Chemical Supplier No. 2932710 MG
An endogenous tricarboxylic acid; formed by the condensation of propionyl-CoA with oxaloacetic acid by citrate synthase under conditions of propionyl-CoA accumulation; accumulation is associated with cobalamin deficiencies, propionic acidemia, and methylmalonic acidurias