Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methylcitric acid trisodium | 117041-96-0 | MFCD16875437 | 96.0% | 272.10 g·mol⁻1 | C7H7Na3O7 | 5 MG
SDP

Catalog No. 5000263348
Encompass_Preferred

2-Methylcitric acid trisodium is the trisodium salt of 2-methylcitric acid provided as a white to off-white solid for laboratory research. It is an endogenous metabolite used as a marker in methylmalonic and propionic acidemias and is employed in biochemical and mitochondrial studies, where it has been reported to inhibit ADP-stimulated mitochondrial respiration supported by glutamate.

  • Trisodium salt, white to off-white solid.
  • CAS number 117041-96-0.
  • Purity 96.0%.
  • Molecular weight 272.10 g·mol⁻1; formula C7H7Na3O7.
  • Highly soluble in water (100 mg/mL; may require ultrasonication).
  • Storage: sealed at 4°C; in solvent -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-63348 Supplier Medchemexpress LLC Supplier No. HY113371A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methylcitric acid trisodium is the trisodium salt of 2-methylcitric acid provided as a white to off-white solid for laboratory research. It is an endogenous metabolite used as a marker in methylmalonic and propionic acidemias and is employed in biochemical and mitochondrial studies, where it has been reported to inhibit ADP-stimulated mitochondrial respiration supported by glutamate.

  • Trisodium salt, white to off-white solid.
  • CAS number 117041-96-0.
  • Purity 96.0%.
  • Molecular weight 272.10 g·mol⁻1; formula C7H7Na3O7.
  • Highly soluble in water (100 mg/mL; may require ultrasonication).
  • Storage: sealed at 4°C; in solvent -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.