Learn More
Medchemexpress LLC 2-Methylcyclohexanone | 583-60-8 | MFCD00001635 | 98.9% | 112.17 | 50 G
SDP
Catalog No. 5000453918
Encompass_Preferred
2-Methylcyclohexanone is a chemical compound primarily used for laboratory research and substance manufacturing. It is a colorless to light yellow liquid with a molecular weight of 112.17 and a purity of 98.9%. This product is for research use only and should be handled by experienced personnel.
  • Consistent with structure
  • Suitable for laboratory research
  • Recommended storage at -20°C for 3 years (pure form) or 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month
  • Can be shipped at room temperature if less than 2 weeks
  • Colorless to light yellow liquid appearance
Catalog No. 50-004-53918 Supplier Medchemexpress LLC Supplier No. HYW01054950G
2-Methylcyclohexanone is a chemical compound primarily used for laboratory research and substance manufacturing. It is a colorless to light yellow liquid with a molecular weight of 112.17 and a purity of 98.9%. This product is for research use only and should be handled by experienced personnel.
  • Consistent with structure
  • Suitable for laboratory research
  • Recommended storage at -20°C for 3 years (pure form) or 4°C for 2 years
  • Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month
  • Can be shipped at room temperature if less than 2 weeks
  • Colorless to light yellow liquid appearance