Learn More
Medchemexpress LLC 2-Methylcyclopentane-1,3-dione | 765-69-5 | 99.9% | 112.13 | 5 G
SDP
Catalog No. 5000451962
Encompass_Preferred
2-Methylcyclopentane-1,3-dione is a key intermediate for the total synthesis of steroids. It is for research use only and not sold to patients.
  • Solid appearance
  • White to light yellow color
  • Initial source: plants Araliaceae Panax ginseng C. A. Meyer
  • Structure classification: ketones, aldehydes, acids
Catalog No. 50-004-51962 Supplier Medchemexpress LLC Supplier No. HYW0129445G
2-Methylcyclopentane-1,3-dione is a key intermediate for the total synthesis of steroids. It is for research use only and not sold to patients.
  • Solid appearance
  • White to light yellow color
  • Initial source: plants Araliaceae Panax ginseng C. A. Meyer
  • Structure classification: ketones, aldehydes, acids