Learn More
Medchemexpress LLC 2-methylcyclopentane-1,3-dione | 765-69-5 | 99.96% | 112.13 | 25 G
SDP
Catalog No. 5000452705
Encompass_Preferred
2-Methylcyclopentane-1,3-dione is a chemical compound that serves as a key intermediate in the total synthesis of steroids. It is also utilized as a drug intermediate, and for general laboratory chemical applications including the manufacture of various substances.
  • Key intermediate for total synthesis of steroids
  • Drug intermediate
  • Suitable for laboratory chemical applications
  • Used in the manufacture of substances
  • White to light yellow solid appearance
  • High purity of 99.96% (HPLC)
Catalog No. 50-004-52705 Supplier Medchemexpress LLC Supplier No. 25GHYW012944
2-Methylcyclopentane-1,3-dione is a chemical compound that serves as a key intermediate in the total synthesis of steroids. It is also utilized as a drug intermediate, and for general laboratory chemical applications including the manufacture of various substances.
  • Key intermediate for total synthesis of steroids
  • Drug intermediate
  • Suitable for laboratory chemical applications
  • Used in the manufacture of substances
  • White to light yellow solid appearance
  • High purity of 99.96% (HPLC)