Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methylcyclopentane-1,3-dione | 765-69-5 | MFCD00001406 | 100.0% | 112.13 | 100 G
SDP

Catalog No. 5000454081
Encompass_Preferred

2-Methylcyclopentane-1,3-dione is a key intermediate for the total synthesis of steroids. It is intended for research use only and not for sale to patients.

  • Key intermediate for total synthesis of steroids
  • For research use only
  • Molecular weight: 112.13
  • Purity: 99.96%
  • CAS number: 765-69-5
  • Appearance: Solid
  • Color: White to light yellow

Catalog No. 50-004-54081 Supplier Medchemexpress LLC Supplier No. HYW012944100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methylcyclopentane-1,3-dione is a key intermediate for the total synthesis of steroids. It is intended for research use only and not for sale to patients.

  • Key intermediate for total synthesis of steroids
  • For research use only
  • Molecular weight: 112.13
  • Purity: 99.96%
  • CAS number: 765-69-5
  • Appearance: Solid
  • Color: White to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.