Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methylheptanoic acid | 1188-02-9 | MFCD00142944 | 99.3% | 144.21 g/mol | C8H16O2 | 1 G
SDP

Catalog No. 5000300747
Encompass_Preferred

2-methylheptanoic acid is a branched-chain carboxylic acid used as a flavouring ingredient and as a research chemical. It is naturally occurring in sources such as honey and is supplied as a high-purity liquid for laboratory and analytical use.

  • High-purity liquid suitable for flavor and research applications.
  • Branched C8 carboxylic acid found in natural products such as honey.
  • Molecular formula C8H16O2; molecular weight 144.21 g/mol.
  • Density approximately 0.906 g/cm³ (typical).
  • Available in small pack sizes for laboratory handling and analysis.
  • Intended for research use; not for human consumption.

Catalog No. 50-003-00747 Supplier Medchemexpress LLC Supplier No. HYW0156641G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-methylheptanoic acid is a branched-chain carboxylic acid used as a flavouring ingredient and as a research chemical. It is naturally occurring in sources such as honey and is supplied as a high-purity liquid for laboratory and analytical use.

  • High-purity liquid suitable for flavor and research applications.
  • Branched C8 carboxylic acid found in natural products such as honey.
  • Molecular formula C8H16O2; molecular weight 144.21 g/mol.
  • Density approximately 0.906 g/cm³ (typical).
  • Available in small pack sizes for laboratory handling and analysis.
  • Intended for research use; not for human consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.