Learn More
Medchemexpress LLC 2-methylheptanoic acid | 1188-02-9 | MFCD00142944 | 99.3% | 144.21 g/mol | C8H16O2 | 10 G
SDP
Catalog No. 5000300674
Encompass_Preferred
2-methylheptanoic acid is a small aliphatic carboxylic acid found in honey and used as a flavoring component and research reagent. It is supplied as a high-purity laboratory chemical, typically stored under low-temperature conditions to preserve stability and purity for analytical or formulation work.
  • Natural flavoring component isolated from honey
  • High purity (99.31%) suitable for research applications
  • Colorless to light yellow liquid with density 0.906 g/cm3
  • Stable under low-temperature storage for extended shelf life
  • Available in small lab-scale packaging for bench use
Catalog No. 50-003-00674 Supplier Medchemexpress LLC Supplier No. HYW01566410G
2-methylheptanoic acid is a small aliphatic carboxylic acid found in honey and used as a flavoring component and research reagent. It is supplied as a high-purity laboratory chemical, typically stored under low-temperature conditions to preserve stability and purity for analytical or formulation work.
  • Natural flavoring component isolated from honey
  • High purity (99.31%) suitable for research applications
  • Colorless to light yellow liquid with density 0.906 g/cm3
  • Stable under low-temperature storage for extended shelf life
  • Available in small lab-scale packaging for bench use