Learn More
Medchemexpress LLC 2-methylheptanoic acid | 1188-02-9 | MFCD00142944 | 99.3% | 144.21 g/mol | C8H16O2 | 100 G
SDP
Catalog No. 5000300297
Encompass_Preferred
2-Methylheptanoic acid is a branched C8 carboxylic acid (CAS 1188-02-9) supplied for research and flavor applications. MedChemExpress lists it as a flavouring ingredient isolated from honey and provides analytical-grade material in liquid form for formulation and study.
  • High chemical purity suitable for research and analytical use.
  • Liquid form with reported density 0.906 g/cm3 for handling and formulation.
  • Molecular weight 144.21 g/mol and formula C8H16O2 for accurate calculations.
  • Used as a flavouring ingredient, isolated from honey, for flavor and fragrance research.
  • Available in laboratory pack sizes up to 100 g for small-scale work.
Catalog No. 50-003-00297 Supplier Medchemexpress LLC Supplier No. HYW015664100G
2-Methylheptanoic acid is a branched C8 carboxylic acid (CAS 1188-02-9) supplied for research and flavor applications. MedChemExpress lists it as a flavouring ingredient isolated from honey and provides analytical-grade material in liquid form for formulation and study.
  • High chemical purity suitable for research and analytical use.
  • Liquid form with reported density 0.906 g/cm3 for handling and formulation.
  • Molecular weight 144.21 g/mol and formula C8H16O2 for accurate calculations.
  • Used as a flavouring ingredient, isolated from honey, for flavor and fragrance research.
  • Available in laboratory pack sizes up to 100 g for small-scale work.