Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methylheptanoic acid | 1188-02-9 | MFCD00142944 | 99.3% | 144.21 g/mol | C8H16O2 | 25 G
SDP

Catalog No. 5000303445
Encompass_Preferred

2-Methylheptanoic acid is a branched C8 carboxylic acid (CAS 1188-02-9) supplied as a high-purity liquid and used as a flavoring ingredient isolated from honey. It is useful for flavor research and small-scale formulation work.

  • Chemical formula C8H16O2.
  • Molecular weight 144.21 g/mol.
  • CAS number 1188-02-9.
  • Purity 99.3%.
  • Physical form liquid; density 0.906 g/cm3.
  • Used as a flavoring ingredient isolated from honey.
  • Available in pack sizes including 25 g.

Catalog No. 50-003-03445 Supplier Medchemexpress LLC Supplier No. 25GHYW015664
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methylheptanoic acid is a branched C8 carboxylic acid (CAS 1188-02-9) supplied as a high-purity liquid and used as a flavoring ingredient isolated from honey. It is useful for flavor research and small-scale formulation work.

  • Chemical formula C8H16O2.
  • Molecular weight 144.21 g/mol.
  • CAS number 1188-02-9.
  • Purity 99.3%.
  • Physical form liquid; density 0.906 g/cm3.
  • Used as a flavoring ingredient isolated from honey.
  • Available in pack sizes including 25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.