Learn More
Medchemexpress LLC 2-methylheptanoic acid | 1188-02-9 | MFCD00142944 | 99.3% | 144.21 g/mol | C8H16O2 | 5 G
SDP
Catalog No. 5000300298
Encompass_Preferred
2-Methylheptanoic acid is a small organic carboxylic acid (CAS 1188-02-9) found in honey and used as a flavoring component and research reagent. Supplied as a high-purity liquid, it is characterized by molecular formula C8H16O2 and molecular weight 144.21 g/mol, and is suitable for flavor, fragrance, and laboratory applications.
  • High purity (99.31%).
  • Liquid physical state with density 0.906 g/cm3.
  • Used as a flavoring ingredient and fragrance intermediate.
  • Suitable as a research reagent and reference standard.
  • Available in multiple pack sizes for laboratory use.
Catalog No. 50-003-00298 Supplier Medchemexpress LLC Supplier No. HYW0156645G
2-Methylheptanoic acid is a small organic carboxylic acid (CAS 1188-02-9) found in honey and used as a flavoring component and research reagent. Supplied as a high-purity liquid, it is characterized by molecular formula C8H16O2 and molecular weight 144.21 g/mol, and is suitable for flavor, fragrance, and laboratory applications.
  • High purity (99.31%).
  • Liquid physical state with density 0.906 g/cm3.
  • Used as a flavoring ingredient and fragrance intermediate.
  • Suitable as a research reagent and reference standard.
  • Available in multiple pack sizes for laboratory use.