Learn More
Medchemexpress LLC 2-methylheptanoic acid | 1188-02-9 | MFCD00142944 | 99.3% | 144.21 g/mol | C8H16O2 | 50 G
SDP
Catalog No. 5000300675
Encompass_Preferred
2-Methylheptanoic acid (CAS 1188-02-9) is a branched aliphatic carboxylic acid used as a flavor and fragrance ingredient and as an intermediate in chemical synthesis. It is a colorless to light-yellow liquid with molecular formula C8H16O2, molecular weight 144.21 g/mol, density 0.906 g/cm3, and reported purity of about 99.3%, suitable for research and formulation work.
  • High purity suitable for research and formulation.
  • Branched C8 carboxylic acid with molecular weight 144.21 g/mol.
  • Colorless to light-yellow liquid with density 0.906 g/cm3.
  • Used as a flavor and fragrance ingredient isolated from natural sources.
  • Useful intermediate for organic synthesis and formulation development.
Catalog No. 50-003-00675 Supplier Medchemexpress LLC Supplier No. HYW01566450G
2-Methylheptanoic acid (CAS 1188-02-9) is a branched aliphatic carboxylic acid used as a flavor and fragrance ingredient and as an intermediate in chemical synthesis. It is a colorless to light-yellow liquid with molecular formula C8H16O2, molecular weight 144.21 g/mol, density 0.906 g/cm3, and reported purity of about 99.3%, suitable for research and formulation work.
  • High purity suitable for research and formulation.
  • Branched C8 carboxylic acid with molecular weight 144.21 g/mol.
  • Colorless to light-yellow liquid with density 0.906 g/cm3.
  • Used as a flavor and fragrance ingredient isolated from natural sources.
  • Useful intermediate for organic synthesis and formulation development.