Learn More
Cayman Chemical 2-MethylhIppurIc AcId 5g
SDP
Catalog No. 502949822
Encompass_Preferred
A metabolite of xylene; has been found in the urine of industrial workers exposed to xylene
Catalog No. 50-294-9822 Supplier Cayman Chemical Supplier No. 343765 G
Add to Cart
Add to Cart
A metabolite of xylene; has been found in the urine of industrial workers exposed to xylene