Learn More
2-Methylimidazole, 97%
Description
2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Methylimidazole is used as a raw material for the preparation of nitroimidazole antibiotics, which is useful in combat anaerobic bacterial and parasitic infections. It acts as a ligand in coordination chemistry. It is also employed as a hardener or accelerator for epoxy resin and auxiliary agent for textile dyes. Further, it acts as a catalyst for refolding of enhanced colored fluorescent protein. It is also useful as a building block in the synthesis of biologically active compounds. In addition to this, it is used as an active pharmaceutical ingredient intermediate such as metronidazole and dimetridazole.
Solubility
Soluble in water and ethanol.
Notes
Incompatible with strong oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications
Specifications
| Melting Point | 142°C to 144°C |
| Boiling Point | 267°C to 268°C |
| Flash Point | 155°C (311°F) |
| Quantity | 2500 g |
| UN Number | UN3259 |
| Beilstein | 1368 |
| Solubility Information | Soluble in water and ethanol. |
| Formula Weight | 82.11 |
| Percent Purity | 97% |
| Physical Form | Crystalline Solid |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H335-H351-H360 |
| Hazard Statement | GHS H StatementH314-H318-H302Causes severe skin burns and eye damage.Causes serious eye damage.Harmful if swallowed. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S. |
| EINECSNumber | 211-765-7 |
| RTECSNumber | NI7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.