Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylindole-3-carboxamide 98.0+%, TCI America™
SDP

Catalog No. C12555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12555G 5 g
C12551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12555G Supplier TCI America Supplier No. C12555G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67242-60-8
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD00143101
InChI Key QKWLFJZXUBMFLU-UHFFFAOYSA-N
Synonym 3-Carbamoyl-2-methylindole
PubChem CID 12415229
IUPAC Name 2-methyl-1H-indole-3-carboxamide
SMILES CC1=C(C2=CC=CC=C2N1)C(=O)N

Specifications

Melting Point 225°C
Color White
Quantity 5 g
Formula Weight 174.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylindole-3-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.