Learn More
Medchemexpress LLC 2-Methylindole | 95-20-5 | 131.17 | 250 G
SDP
Catalog No. 5000449921
Encompass_Preferred
2-Methylindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is utilized in various synthetic processes and is a versatile tool for chemical synthesis and biological studies.
  • Regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction
  • Friedel-Crafts alkylation reactions
  • Preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Michael addition reactions
  • Synthesis of cyclooxygenase-1 (COX-1) and cyclooxygenase-2 (COX-2) inhibitors
Catalog No. 50-004-49921 Supplier Medchemexpress LLC Supplier No. 250GHYW007556
2-Methylindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is utilized in various synthetic processes and is a versatile tool for chemical synthesis and biological studies.
  • Regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction
  • Friedel-Crafts alkylation reactions
  • Preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Michael addition reactions
  • Synthesis of cyclooxygenase-1 (COX-1) and cyclooxygenase-2 (COX-2) inhibitors