Learn More
Medchemexpress LLC 2-Methylindole | 95-20-5 | 131.17 | 500 G
SDP
Catalog No. 5000445965
Encompass_Preferred
2-Methylindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction.
  • Friedel-Crafts alkylation reactions.
  • Preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.
  • Michael addition reactions.
  • Synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.
Catalog No. 50-004-45965 Supplier Medchemexpress LLC Supplier No. HYW007556500G
2-Methylindole is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction.
  • Friedel-Crafts alkylation reactions.
  • Preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.
  • Michael addition reactions.
  • Synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.