Learn More
Medchemexpress LLC 2-Methylindole | 95-20-5 | 131.18 | 25 G
SDP
Catalog No. 5000449851
Encompass_Preferred
2-Methylindole is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is suitable for various synthetic applications.
  • Reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction.
  • Used for Friedel-Crafts alkylation reactions.
  • Preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.
  • Used for Michael addition reactions.
  • Synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.
Catalog No. 50-004-49851 Supplier Medchemexpress LLC Supplier No. 25GHYW007556
2-Methylindole is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is suitable for various synthetic applications.
  • Reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction.
  • Used for Friedel-Crafts alkylation reactions.
  • Preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.
  • Used for Michael addition reactions.
  • Synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.