Learn More
Medchemexpress LLC 2-Methylindole | 95-20-5 | MFCD00005616 | 131.17 | 1000 G
SDP
Catalog No. 5000445813
Encompass_Preferred
2-Methylindole is a biochemical reagent suitable for use as a biological material or organic compound in life science research. It functions as a reactant in various synthetic processes and is for research use only.
  • Regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction.
  • Friedel-Crafts alkylation reactions.
  • Preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential Anticancer immunomodulators.
  • Michael addition reactions.
  • Synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.
Catalog No. 50-004-45813 Supplier Medchemexpress LLC Supplier No. HYW0075561000G
2-Methylindole is a biochemical reagent suitable for use as a biological material or organic compound in life science research. It functions as a reactant in various synthetic processes and is for research use only.
  • Regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction.
  • Friedel-Crafts alkylation reactions.
  • Preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential Anticancer immunomodulators.
  • Michael addition reactions.
  • Synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.