Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methylisoborneol Solution

Catalog No. M39331ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M39331ML 25 g
M39335X1ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. M39331ML Supplier Sigma Aldrich Supplier No. M39331ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methylisoborneol (2-MIB) is a semi-volatile compound, responsible for the undesirable earthy-musty odor in surface water. It can be efficiently degraded by TiO2 photocatalysis. 2-MIB can also be easily adsorbed on granular activated carbon.2-Methylisoborneol can be used as a reactant: To prepare N-[(1R,2R,4R)-1,4,7,7-tetramethylbicyclo[2.2.1]hept-2-yl]acetamide by reacting with acetonitrile in the presence of boron trifluoride etherate. In the enantioselective total synthesis of geissoschizine as well as geissoschizol.Fungal metabolite which contributes to the earthy odor in public water supplies.

Specifications

CAS 592-85-8
Melting Point 165°C
Density 3.71 g/mL at 25°C
Molecular Formula C2HgN2S2
Linear Formula Hg(SCN)2
MDL Number MFCD00011039
Quantity 25 g
Synonym (1R-exo)-1,2,7,7-Tetramethylbicyclo[2.2.1]heptan-2-ol
Molecular Weight (g/mol) 316.75 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.