Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methylisoborneol Solution

Catalog No. M39335X1ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M39335X1ML 1 mL
M39331ML 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. M39335X1ML Supplier Sigma Aldrich Supplier No. M39335X1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methylisoborneol (2-MIB) is a semi-volatile compound, responsible for the undesirable earthy-musty odor in surface water. It can be efficiently degraded by TiO2 photocatalysis. 2-MIB can also be easily adsorbed on granular activated carbon.2-Methylisoborneol can be used as a reactant: To prepare N-[(1R,2R,4R)-1,4,7,7-tetramethylbicyclo[2.2.1]hept-2-yl]acetamide by reacting with acetonitrile in the presence of boron trifluoride etherate. In the enantioselective total synthesis of geissoschizine as well as geissoschizol.Fungal metabolite which contributes to the earthy odor in public water supplies.

Specifications

CAS 2371-42-8
Molecular Formula C11H20O
Linear Formula C11H20O
MDL Number MFCD00082463
Quantity 1 mL
Synonym (1R-exo)-1,2,7,7-Tetramethylbicyclo[2.2.1]heptan-2-ol
Molecular Weight (g/mol) 168.28 g/mol
Biological Activity Eyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.