Learn More
FUJIFILM Biosciences 2-Methylisoborneol Standard
Catalog No. 5000450364
Encompass_Preferred
This product is a high-purity standard of 2-Methylisoborneol commonly used for water analysis applications via capillary gas chromatography 2-MIB is one source of the earthy-musty taste and odor in water These problems have been reported when levels are as little as 10-15 parts per trillion 2-MIB Standard is synthetically produced as a single isomer Provided in crystalline form with a minimum purity of 98% it is soluble in organic solvents and slightly soluble in water Stored in a glass container under inert gas to maintain stability
Catalog No. 50-004-50364 Supplier FUJIFILM Biosciences Supplier No. 13207071
This product is a high-purity standard of 2-Methylisoborneol commonly used for water analysis applications via capillary gas chromatography 2-MIB is one source of the earthy-musty taste and odor in water These problems have been reported when levels are as little as 10-15 parts per trillion 2-MIB Standard is synthetically produced as a single isomer Provided in crystalline form with a minimum purity of 98% it is soluble in organic solvents and slightly soluble in water Stored in a glass container under inert gas to maintain stability