Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylisothiazolone Hydrochloride, ≥99.9, MP Biomedicals

Catalog No. ICN15952290 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15952290 500 mg
ICN15952291 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15952290 Supplier MP Biomedicals, Inc Supplier No. 0215952290
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26172-54-3
Molecular Formula C4H6ClNOS
Molecular Weight (g/mol) 151.608
MDL Number MFCD06804636
InChI Key SJXPQSRCFCPWQQ-UHFFFAOYSA-N
Synonym 2-Methyl-4-isothiazoline-3-one, MIT
PubChem CID 117746
IUPAC Name 2-methyl-1,2-thiazol-3-one;hydrochloride
SMILES CN1C(=O)C=CS1.Cl

Specifications

Type 2-Methylisothiazolone Hydrochloride
Physical Form Crystalline
Color White
Quantity 500 mg
Assay Percent Range ≥99%
Formula Weight 151.6
Chemical Name or Material 2-Methylisothiazolone Hydrochloride
EINECSNumber 247-499-3
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.